Gene Transcripts

Gene Symbol
CYLC2
Entrez Gene ID
1539
Gene Name
cylicin 2
Gene Aliases
Chromosome Location
Chr. 9: 102995311 - 103018488 on Build GRCh38
UniGene ID
Hs.3232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001340.4 NP_001331.1 7 1994
GenBank mRNA BX097520.1 - 3 430
Z46788.1 - 7 1924

View Full Product Details