Gene Transcripts
- Gene Symbol
- Slc7a15
- Entrez Gene ID
- 328059
- Gene Name
- solute carrier family 7 (cationic amino acid transporter, y+ system), member 15
- Gene Aliases
- Arpat
- Chromosome Location
- Chr. 12: 8528481 - 8599066 on Build GRCm38
- UniGene ID
- Mm.46326
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033481.1 |
- |
7 |
1539 |
|
View Full Product Details