Gene Transcripts

Gene Symbol
Slc30a9
Entrez Gene ID
109108
Gene Name
solute carrier family 30 (zinc transporter), member 9
Gene Aliases
2310024J23Rik, AL024256, GAC63, HUEL
Chromosome Location
Chr. 5: 67306937 - 67356834 on Build GRCm38
UniGene ID
Mm.234455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178651.3 NP_848766.2 18 3056
GenBank mRNA AK033990.1 - 2081
AK082853.1 - 18 3058

View Full Product Details