Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QSOX1
Entrez Gene ID
838067
Gene Name
quiescin-sulfhydryl oxidase 1
Gene Aliases
AT1G15020, ATQSOX1, QSO2, T15D22.7, T15D22_7, quiescin-sulfhydryl oxidase 1
Chromosome Location
Chr.1: 5172232 - 5176479 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332143.1 NP_001321934.1 4 - 5 837 145
NM_101371.5 NP_172955.1 4 - 5 887 145
NM_179333.2 NP_849664.1 4 - 5 837 145
GenBank mRNA AY062528.1 - 4 - 5 628 145
BT001218.1 - 4 - 5 513 145

View Full Product Details