Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCG35
- Entrez Gene ID
-
838087
- Gene Name
- pleiotropic drug resistance 7
- Gene Aliases
-
AT1G15210, ATP-binding cassette G35, ATPDR7, F9L1.15, F9L1_15, PDR7, PLEIOTROPIC DRUG RESISTANCE 7, pleiotropic drug resistance 7
- Chromosome Location
-
Chr.1: 5231345 - 5236886 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101389.3 |
NP_172973.1 |
3 - 4 |
1662 |
119 |
View Full Product Details