Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBD10
Entrez Gene ID
838103
Gene Name
methyl-CPG-binding domain 10
Gene Aliases
AT1G15340, F9L1.28, F9L1_28, methyl-CPG-binding domain 10
Chromosome Location
Chr.1: 5275616 - 5277869 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198074.1 NP_001185003.1 1 - 2 273 73
NM_101403.4 NP_563971.1 1 - 2 453 73
GenBank mRNA AK230256.1 - - 1209 73
AY094439.1 - 1 - 2 206 73
BT020367.1 - 1 - 2 58 73

View Full Product Details