Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDF1A
Entrez Gene ID
838109
Gene Name
peptide deformylase 1A
Gene Aliases
AT1G15390, ATDEF1, peptide deformylase 1A
Chromosome Location
Chr.1: 5294629 - 5295931 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101408.3 NP_563974.1 2 - 3 525 99
GenBank mRNA AF250959.1 - 2 - 3 525 99

View Full Product Details