Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOS11
Entrez Gene ID
838158
Gene Name
golgi snare 11
Gene Aliases
AT1G15880, F7H2.21, F7H2_21, atgos11, golgi snare 11
Chromosome Location
Chr.1: 5458398 - 5460359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101456.4 NP_563985.1 4 - 5 833 92
GenBank mRNA AF332426.1 - 4 - 5 563 92
AF357528.1 - 4 - 5 813 92
AF410263.1 - 4 - 5 805 92
AY093723.1 - 4 - 5 563 92

View Full Product Details