Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WAKL6
- Entrez Gene ID
-
838180
- Gene Name
- wall associated kinase-like 6
- Gene Aliases
-
AT1G16110, T24D18.30, T24D18_30, wall associated kinase-like 6
- Chromosome Location
-
Chr.1: 5518219 - 5520929 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101478.3 |
NP_173062.1 |
1 - 2 |
1120 |
83 |
View Full Product Details