Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WAKL1
- Entrez Gene ID
-
838181
- Gene Name
- wall associated kinase-like 1
- Gene Aliases
-
AT1G16120, T24D18.20, T24D18_20, wall associated kinase-like 1
- Chromosome Location
-
Chr.1: 5522602 - 5525452 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101479.2 |
NP_173063.1 |
1 - 2 |
979 |
72 |
View Full Product Details