Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAKL2
Entrez Gene ID
838182
Gene Name
wall associated kinase-like 2
Gene Aliases
AT1G16130, T24D18.21, T24D18_21, wall associated kinase-like 2
Chromosome Location
Chr.1: 5525293 - 5528212 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101480.3 NP_173064.1 1 - 2 1251 99
GenBank mRNA AK229684.1 - 1 - 2 1053 99
BT008569.1 - 1 - 2 910 99
BT008657.1 - 1 - 2 1053 99

View Full Product Details