Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAPCP-2
Entrez Gene ID
838199
Gene Name
glyceraldehyde-3-phosphate dehydrogenase of plastid 2
Gene Aliases
AT1G16300, F3O9.10, F3O9_10, glyceraldehyde-3-phosphate dehydrogenase of plastid 2
Chromosome Location
Chr.1: 5574257 - 5577622 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101496.3 NP_173080.1 2 - 3 343 82
GenBank mRNA BT021096.1 - 2 - 3 223 82

View Full Product Details