Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCB3;1
Entrez Gene ID
838202
Gene Name
cyclin b3;1
Gene Aliases
AT1G16330, F3O9.13, F3O9_13, cyclin b3;1
Chromosome Location
Chr.1: 5582387 - 5587461 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101499.4 NP_173083.3 18 - 19 1376 115

View Full Product Details