Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OCT6
- Entrez Gene ID
-
838207
- Gene Name
- organic cation/carnitine transporter 6
- Gene Aliases
-
ARABIDOPSIS THALIANA ORGANIC CATION/CARNITINE TRANSPORTER 6, AT1G16370, ATOCT6, F3O9.17, F3O9_17, organic cation/carnitine transporter 6
- Chromosome Location
-
Chr.1: 5596633 - 5598447 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101503.4 |
NP_173087.1 |
1 - 1 |
1673 |
72 |
GenBank mRNA |
AK229354.1 |
- |
1 - 1 |
1613 |
72 |
View Full Product Details