Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS2
Entrez Gene ID
838269
Gene Name
trehalose-6-phosphate synthase
Gene Aliases
AT1G16980, ATTPS2, F6I1.1, F6I1_1, TREHALOSE -6-PHOSPHATASE SYNTHASE S2, trehalose-phosphatase/synthase 2
Chromosome Location
Chr.1: 5807200 - 5811610 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332273.1 NP_001321461.1 11 - 12 1651 101
NM_101559.2 NP_173142.2 11 - 12 1651 101

View Full Product Details