Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TPS3
- Entrez Gene ID
-
838270
- Gene Name
- trehalose-phosphatase/synthase 3
- Gene Aliases
-
AT1G17000, ATTPS3, F20D23.30, F20D23_30, TREHALOSE -6-PHOSPHATASE SYNTHASE S3, trehalose-phosphatase/synthase 3
- Chromosome Location
-
Chr.1: 5812728 - 5816662 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101560.5 |
NP_173143.4 |
3 - 4 |
562 |
82 |
View Full Product Details