Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD7
Entrez Gene ID
838321
Gene Name
IQ-domain 7
Gene Aliases
AT1G17480, F28G4.3, F28G4_3, IQ-domain 7
Chromosome Location
Chr.1: 6006413 - 6008501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101610.3 NP_173191.2 2 - 3 689 68
GenBank mRNA AY702665.1 - 2 - 3 320 68
BT032868.1 - 2 - 3 320 68

View Full Product Details