Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYA1
Entrez Gene ID
838333
Gene Name
myosin 1
Gene Aliases
ARABIDOPSIS MYOSIN 1, AT1G17580, ATMYA1, F1L3.28, F1L3_28, MYOSIN XI 1, XI-1, myosin 1
Chromosome Location
Chr.1: 6039250 - 6049685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101620.3 NP_173201.2 34 - 35 4367 80
GenBank mRNA Z28389.1 - - 4314 80

View Full Product Details