Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GLYR2
- Entrez Gene ID
-
838342
- Gene Name
- glyoxylate reductase 2
- Gene Aliases
-
AT1G17650, AtGLYR2, F11A6.12, GLYOXYLATE REDUCTASE 2, GR2, glyoxylate reductase 2
- Chromosome Location
-
Chr.1: 6069424 - 6072140 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101628.4 |
NP_564030.2 |
1 - 2 |
354 |
69 |
View Full Product Details