Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CSTF77
- Entrez Gene ID
-
838354
- Gene Name
- Tetratricopeptide repeat (TPR)-like superfamily protein
- Gene Aliases
-
ARABIDOPSIS THALIANA CLEAVAGE STIMULATION FACTOR 77, AT1G17760, ATCSTF77, F11A6.10, F11A6_10
- Chromosome Location
-
Chr.1: 6110107 - 6116617 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101639.3 |
NP_173218.2 |
18 - 19 |
1941 |
150 |
View Full Product Details