Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EGY3
- Entrez Gene ID
-
838366
- Gene Name
- ethylene-dependent gravitropism-deficient and yellow-green-like 3
- Gene Aliases
-
AT1G17870, ATEGY3, ETHYLENE-DEPENDENT GRAVITROPISM-DEFICIENT AND YELLOW-GREEN-LIKE 3, F2H15.10, F2H15_10
- Chromosome Location
-
Chr.1: 6149939 - 6152222 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101650.3 |
NP_173229.1 |
1 - 2 |
586 |
131 |
View Full Product Details