Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG12
Entrez Gene ID
838371
Gene Name
homeodomain GLABROUS 12
Gene Aliases
AT1G17920, F2H15.14, F2H15_14, homeodomain GLABROUS 12
Chromosome Location
Chr.1: 6161870 - 6165820 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332324.1 NP_001323262.1 7 - 8 1700 82
NM_101655.3 NP_564041.2 6 - 7 1968 82
GenBank mRNA AB493465.1 - 6 - 7 1181 82

View Full Product Details