Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DA1
Entrez Gene ID
838510
Gene Name
DA1
Gene Aliases
AT1G19270, DA (large in Chinese) 1, T29M8.14, T29M8_14
Chromosome Location
Chr.1: 6662417 - 6666032 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332389.1 NP_001320535.1 7 - 8 1162 135
NM_101785.4 NP_173361.1 6 - 7 1355 135

View Full Product Details