Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARVUS
Entrez Gene ID
838512
Gene Name
Nucleotide-diphospho-sugar transferases superfamily protein
Gene Aliases
AT1G19300, ATGATL1, F18O14.2, F18O14_2, GALACTURONOSYLTRANSFERASE-LIKE 1, GAOLAOZHUANGREN 1, GATL1, GLZ1
Chromosome Location
Chr.1: 6671095 - 6672777 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101787.3 NP_564077.1 1 - 1 1471 152
GenBank mRNA AF424600.1 - 1 - 1 1266 153

View Full Product Details