Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATTRX4
Entrez Gene ID
838562
Gene Name
Thioredoxin superfamily protein
Gene Aliases
AT1G19730, ATH4, F14P1.32, F14P1_32, THIOREDOXIN, thioredoxin H-type 4
Chromosome Location
Chr.1: 6822847 - 6824071 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101829.4 NP_173403.1 2 - 3 267 74
GenBank mRNA AK118542.1 - 2 - 3 238 74
BT004710.1 - 2 - 3 216 74

View Full Product Details