Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd32
Entrez Gene ID
838575
Gene Name
IQ-domain 32
Gene Aliases
AT1G19870, F6F9.8, F6F9_8, IQ-domain 32, IQD32
Chromosome Location
Chr.1: 6895009 - 6898818 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101842.5 NP_564097.1 1 - 2 373 69

View Full Product Details