Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC5B
Entrez Gene ID
838713
Gene Name
Exocyst complex component SEC5
Gene Aliases
AT1G21170
Chromosome Location
Chr.1: 7412830 - 7419643 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332488.1 NP_001322652.1 15 - 16 2487 143
NM_101971.3 NP_173541.1 15 - 16 2680 143

View Full Product Details