Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9C1
Entrez Gene ID
841315
Gene Name
glycosyl hydrolase 9C1
Gene Aliases
AT1G48930, AtGH9C1, F27K7.5, glycosyl hydrolase 9C1
Chromosome Location
Chr.1: 18101643 - 18104768 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103786.3 NP_175323.1 2 - 3 809 88
GenBank mRNA BT002935.1 - 2 - 3 702 88
BT005638.1 - 2 - 3 628 88

View Full Product Details