Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHI
Entrez Gene ID
836767
Gene Name
Lateral root primordium (LRP) protein-like protein
Gene Aliases
AT5G66350, K1L20.13, K1L20_13, SHORT INTERNODES
Chromosome Location
Chr.5: 26503115 - 26506809 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345758.1 NP_001330898.1 1 - 2 1291 85
NM_001345759.1 NP_001330899.1 1 - 2 1080 85
NM_126033.2 NP_201436.1 1 - 2 1291 85
GenBank mRNA AB493816.1 - 1 - 2 625 85
AF152555.1 - 1 - 2 1051 85

View Full Product Details