Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks17
Entrez Gene ID
836826
Gene Name
SKU5 similar 17
Gene Aliases
AT5G66920, MUD21.18, MUD21_18, SKU5 similar 17, SKU5 similar 17
Chromosome Location
Chr.5: 26722701 - 26725657 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126091.4 NP_569041.1 5 - 6 1618 69
GenBank mRNA AY094401.1 - 5 - 6 1469 69
BT000862.1 - 5 - 6 1356 69

View Full Product Details