Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCD3;2
Entrez Gene ID
836861
Gene Name
CYCLIN D3;2
Gene Aliases
AT5G67260, CYCLIN D3;2, K3G17.2, K3G17_2
Chromosome Location
Chr.5: 26836139 - 26839597 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345820.1 NP_001331659.1 3 - 4 1008 98
NM_126126.3 NP_201527.1 3 - 4 1009 98
GenBank mRNA AY062645.1 - 3 - 4 956 98
AY087540.1 - 3 - 4 1000 98
AY093254.1 - 3 - 4 835 98

View Full Product Details