Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOS15
Entrez Gene ID
836867
Gene Name
WD-40 repeat family protein
Gene Aliases
AT5G67320, K8K14.4, K8K14_4, high expression of osmotically responsive genes 15
Chromosome Location
Chr.5: 26857154 - 26861119 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126132.4 NP_201533.1 4 - 5 914 99
GenBank mRNA AY057698.1 - 4 - 5 914 99
AY143932.1 - 4 - 5 800 99

View Full Product Details