Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRO1
Entrez Gene ID
836895
Gene Name
NADH-ubiquinone oxidoreductase-like protein
Gene Aliases
AT5G67590, FROSTBITE1, K9I9.16, K9I9_16
Chromosome Location
Chr.5: 26957915 - 26959557 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126159.4 NP_201560.1 4 - 5 570 69
GenBank mRNA AK118663.1 - 4 - 5 461 69
BT003675.1 - 4 - 5 412 69

View Full Product Details