Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAA32
Entrez Gene ID
814648
Gene Name
indole-3-acetic acid inducible 32
Gene Aliases
AT2G01200, F10A8.8, F10A8_8, MEE10, indole-3-acetic acid inducible 32, maternal effect embryo arrest 10
Chromosome Location
Chr.2: 117969 - 120095 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335044.1 NP_001318172.1 3 - 4 923 69
NM_201661.2 NP_973390.1 4 - 5 680 69
GenBank mRNA AY669803.1 - 3 - 4 507 69
DQ446449.1 - 3 - 4 507 69

View Full Product Details