Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEE65
- Entrez Gene ID
-
814656
- Gene Name
- Cyclin/Brf1-like TBP-binding protein
- Gene Aliases
-
AT2G01280, F10A8.16, F10A8_16, maternal effect embryo arrest 65
- Chromosome Location
-
Chr.2: 145676 - 149394 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_126189.4 |
NP_178237.3 |
16 - 17 |
1470 |
62 |
View Full Product Details