Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QPT
Entrez Gene ID
814663
Gene Name
quinolinate phoshoribosyltransferase
Gene Aliases
AT2G01350, F10A8.23, F10A8_23, quinolinate phoshoribosyltransferase
Chromosome Location
Chr.2: 165141 - 167259 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084395.1 NP_001077864.1 6 - 7 731 117
NM_001084396.1 NP_001077865.1 5 - 6 831 117
NM_126196.3 NP_565259.1 6 - 7 755 117
NM_201664.2 NP_973393.1 5 - 6 821 117
GenBank mRNA AK117875.1 - 6 - 7 725 117
AY057685.1 - 6 - 7 727 117
AY116955.1 - 6 - 7 705 117

View Full Product Details