Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE11
Entrez Gene ID
814691
Gene Name
RNI-like superfamily protein
Gene Aliases
AT2G01620, T8O11.21, T8O11_21, maternal effect embryo arrest 11
Chromosome Location
Chr.2: 278124 - 279296 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126223.3 NP_565268.1 2 - 3 655 65
GenBank mRNA AK117653.1 - 2 - 3 652 65
BT005175.1 - 2 - 3 575 65

View Full Product Details