Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXGT-A3
Entrez Gene ID
814716
Gene Name
endoxyloglucan transferase A3
Gene Aliases
AT2G01850, ATXTH27, T23K3.4, T23K3_4, XTH27, XYLOGLUCAN ENDOTRANSGLUCOSYLASE/HYDROLASE 27, endoxyloglucan transferase A3
Chromosome Location
Chr.2: 385171 - 387478 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126246.4 NP_178294.1 3 - 4 692 147
GenBank mRNA AY059910.1 - 3 - 4 640 147
BT008820.1 - 3 - 4 489 147
D63509.1 - 3 - 4 628 147

View Full Product Details