Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP8
Entrez Gene ID
814720
Gene Name
purple acid phosphatase 8
Gene Aliases
AT2G01890, ATPAP8, PURPLE ACID PHOSPHATASE 8, T23K3.8, T23K3_8, purple acid phosphatase 8
Chromosome Location
Chr.2: 396756 - 399338 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335083.1 NP_001325278.1 5 - 6 837 104
NM_126250.5 NP_178298.2 6 - 7 960 104
NM_201668.2 NP_973397.1 6 - 7 746 104
GenBank mRNA AF492660.1 - 6 - 7 790 104
AY065434.1 - 6 - 7 830 104
AY117232.1 - 6 - 7 790 104

View Full Product Details