Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TET14
Entrez Gene ID
814727
Gene Name
tetraspanin14
Gene Aliases
AT2G01960, F14H20.3, F14H20_3, tetraspanin14
Chromosome Location
Chr.2: 450379 - 451910 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126257.3 NP_178305.2 1 - 2 515 87
GenBank mRNA AY954776.1 - 1 - 2 501 87

View Full Product Details