Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR72
Entrez Gene ID
814745
Gene Name
low-molecular-weight cysteine-rich 72
Gene Aliases
AT2G02140, F5O4.9, F5O4_9, PDF2.6, low-molecular-weight cysteine-rich 72
Chromosome Location
Chr.2: 544647 - 545564 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126274.3 NP_178322.1 1 - 2 173 72
GenBank mRNA AY087062.1 - 1 - 2 113 72
BT028882.1 - 1 - 2 65 72

View Full Product Details