Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOM3
Entrez Gene ID
814749
Gene Name
tobamovirus multiplication protein 3
Gene Aliases
AT2G02180, F5O4.5, F5O4_5, tobamovirus multiplication protein 3
Chromosome Location
Chr.2: 560695 - 563367 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126278.3 NP_027422.1 3 - 4 616 85
GenBank mRNA AB036427.1 - 3 - 4 354 85
AK117222.1 - 3 - 4 383 85

View Full Product Details