Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-B8
Entrez Gene ID
814765
Gene Name
phloem protein 2-B8
Gene Aliases
AT2G02340, AtPP2-B8, T16F16.13, T16F16_13, phloem protein 2-B8
Chromosome Location
Chr.2: 616088 - 617322 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126291.2 NP_178339.1 1 - 2 396 125

View Full Product Details