Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTR1
Entrez Gene ID
814811
Gene Name
UDP-galactose transporter 1
Gene Aliases
AT2G02810, ATUTR1, T20F6.5, T20F6_5, UDP-GALACTOSE TRANSPORTER 1, UDP-galactose transporter 1
Chromosome Location
Chr.2: 801527 - 803521 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126336.3 NP_565290.1 3 - 4 447 135
GenBank mRNA AF370496.1 - 3 - 4 446 135
BT020431.1 - 3 - 4 331 135

View Full Product Details