Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARPN
Entrez Gene ID
814816
Gene Name
plantacyanin
Gene Aliases
AT2G02850, T17M13.2, T17M13_2, plantacyanin
Chromosome Location
Chr.2: 826299 - 827994 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126340.3 NP_178388.1 1 - 2 455 75
GenBank mRNA AF325063.1 - 1 - 2 181 75
AY064141.1 - 1 - 2 242 75
AY097407.1 - 1 - 2 181 75

View Full Product Details