Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPP
Entrez Gene ID
814841
Gene Name
signal peptide peptidase
Gene Aliases
AT2G03120, ATSPP, T18E12.21, T18E12_21, signal peptide peptidase
Chromosome Location
Chr.2: 937383 - 940551 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126363.7 NP_565294.1 8 - 9 899 114
GenBank mRNA AY065169.1 - 8 - 9 861 114
AY114571.1 - 8 - 9 728 114

View Full Product Details