The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001125078.1 | NP_001118550.1 | 1 - 2 | 328 | 116 |
NM_111014.4 | NP_186797.1 | 1 - 2 | 391 | 116 | |
GenBank mRNA | AY039843.1 | - | 1 - 2 | 326 | 116 |
AY087781.1 | - | 1 - 2 | 283 | 116 | |
AY113168.1 | - | 1 - 2 | 143 | 116 | |
AY568524.1 | - | 1 - 2 | 143 | 116 |