Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP38
Entrez Gene ID
821137
Gene Name
cyclophilin 38
Gene Aliases
ARABIDOPSIS CYCLOPHILIN 38, AT3G01480, ATCYP38, F4P13.3, F4P13_3, cyclophilin 38
Chromosome Location
Chr.3: 188321 - 190892 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125078.1 NP_001118550.1 3 - 4 818 88
NM_111014.4 NP_186797.1 3 - 4 881 88
GenBank mRNA AY039843.1 - 3 - 4 816 88
AY087781.1 - 3 - 4 773 88
AY113168.1 - 3 - 4 633 88
AY568524.1 - 3 - 4 633 88

View Full Product Details