Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGLG1
Entrez Gene ID
819911
Gene Name
RING domain ligase1
Gene Aliases
AT3G01650, F4P13.19, F4P13_19, RING domain ligase1
Chromosome Location
Chr.3: 241739 - 245284 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337352.1 NP_001325948.1 3 - 4 1392 108
NM_111031.4 NP_186814.1 3 - 4 894 108

View Full Product Details