Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACR6
Entrez Gene ID
821217
Gene Name
ACT domain repeat 6
Gene Aliases
ACT domain repeat 6, AT3G01990, F1C9.23, F1C9_23
Chromosome Location
Chr.3: 330099 - 333132 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337368.1 NP_001319446.1 5 - 6 1092 69
NM_001337369.1 NP_001327460.1 5 - 6 1092 69
NM_001337371.1 NP_001327454.1 4 - 5 1080 69
NM_001337373.1 NP_001327458.1 5 - 6 1015 69
NM_001337374.1 NP_001327457.1 5 - 6 1015 69
NM_001337375.1 NP_001327456.1 5 - 6 1015 69
NM_001337376.1 NP_001327453.1 4 - 5 1157 69
NM_111065.3 NP_186848.1 5 - 6 1092 69
GenBank mRNA AF528062.1 - 5 - 6 1092 69
AK118222.1 - 5 - 6 1044 69
BT005556.1 - 5 - 6 973 69

View Full Product Details